Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172844.2 | NP_766432.1 | 10 | 2029 | |
GenBank mRNA | AK029219.1 | - | 10 | 2030 | |
AK161224.1 | - | 10 | 2029 | ||
BC111885.1 | - | 8 | 1758 | 40060432 | |
BC112405.1 | - | 10 | 2019 | 40060428 |