Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199283.1 | NP_001186212.1 | 14 | 3785 | |
NM_001199284.1 | NP_001186213.1 | 14 | 3567 | ||
NM_173388.2 | NP_775564.1 | 14 | 3530 | ||
GenBank mRNA | AK143773.1 | - | 14 | 3628 | |
AK158440.1 | - | 5 | 2511 | ||
BC042513.1 | - | 14 | 3526 | 3964020 |