Gene Transcripts

Gene Symbol
Oog4
Entrez Gene ID
242737
Gene Name
oogenesin 4
Gene Aliases
E330017M15
Chromosome Location
Chr. 4: 143437164 - 143450308 on Build GRCm38
UniGene ID
Mm.148643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173773.1 NP_776134.1 3 462
GenBank mRNA AK054346.1 - 3 484
AY573564.1 - 3 462
BC053722.1 - 2 333
BC098485.1 - 2 321 30051767

View Full Product Details