Gene Transcripts
- Gene Symbol
- ENTPD3
- Entrez Gene ID
- 956
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 3
- Gene Aliases
- CD39L3, HB6, NTPDase-3
- Chromosome Location
- Chr. 3: 40387156 - 40428626 on Build GRCh38
- UniGene ID
- Hs.441145
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001248.3 |
NP_001239.2
|
4, 5 |
412 |
|
NM_001291960.1 |
NP_001278889.1
|
4, 5 |
453 |
|
NM_001291961.1 |
NP_001278890.1
|
4, 5 |
412 |
|
XM_011534266.2 |
XP_011532568.1
|
4, 5 |
412 |
|
GenBank mRNA |
AF034840.2 |
- |
4, 5 |
373 |
|
AF039917.1 |
- |
4, 5 |
376 |
|
AK313322.1 |
- |
4, 5 |
438 |
|
BC029869.1 |
- |
3, 4 |
316 |
5174918 |
BX952692.1 |
- |
4, 5 |
387 |
|
DA209533.1 |
- |
4, 5 |
412 |
|
View Full Product Details