Gene Transcripts

Gene Symbol
Neu4
Entrez Gene ID
241159
Gene Name
sialidase 4
Gene Aliases
9330166I04
Chromosome Location
Chr. 1: 94020446 - 94028334 on Build GRCm38
UniGene ID
Mm.214565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173772.3 NP_776133.1 4 902
GenBank mRNA AB510404.1 - 5 883
AK034236.1 - 4 873
AY258421.1 - 4 815
BC070438.1 - 4 902 6842155

View Full Product Details