Gene Transcripts

Gene Symbol
Als2cr11
Entrez Gene ID
73463
Gene Name
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human)
Gene Aliases
1700052H20Rik, 4930408G06Rik
Chromosome Location
Chr. 1: 59050506 - 59095015 on Build GRCm38
UniGene ID
Mm.252811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175200.4 NP_780409.2 6 826
GenBank mRNA AK076617.1 - 6 828
BC118929.2 - 6 792 40090362

View Full Product Details