Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293715.1 | NP_001280644.1 | 2 | 584 | |
NM_175356.3 | NP_780565.2 | 2 | 584 | ||
GenBank mRNA | AK028974.1 | - | 2 | 569 | |
AK030704.1 | - | 2 | 344 | ||
AK053614.1 | - | 2 | 579 | ||
AK157918.1 | - | 2 | 376 | ||
AK172074.1 | - | 2 | 577 | ||
BC079846.1 | - | 2 | 584 | 5707855 | |
BC113176.1 | - | 2 | 439 | 40060773 | |
BC138457.1 | - | 2 | 487 | 8861477 | |
BC138458.1 | - | 2 | 487 | 8861478 | |
BQ443265.1 | - | 2 | 584 |