Gene Transcripts

Gene Symbol
Tprn
Entrez Gene ID
97031
Gene Name
taperin
Gene Aliases
C430004E15Rik, C87750
Chromosome Location
Chr. 2: 25262598 - 25269886 on Build GRCm38
UniGene ID
Mm.308539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175286.4 NP_780495.2 1 950
GenBank mRNA AK082888.1 - 1 931
AK165772.1 - 1 892
AK171799.1 - 1 964
BC089508.1 - 1 460 6589951

View Full Product Details