Gene Transcripts

Gene Symbol
Fbxo48
Entrez Gene ID
319701
Gene Name
F-box protein 48
Gene Aliases
A630050E13Rik
Chromosome Location
Chr. 11: 16951375 - 16954777 on Build GRCm38
UniGene ID
Mm.120672
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176982.2 NP_795956.1 3 1381
GenBank mRNA AK037835.1 - 3 1381
AK041978.1 - 3 688
BC125589.1 - 2 258 40130104
BC125591.1 - 2 258 40130106
BC145147.1 - 1 353

View Full Product Details