Gene Transcripts
- Gene Symbol
- FABP5
- Entrez Gene ID
- 2171
- Gene Name
- fatty acid binding protein 5
- Gene Aliases
- E-FABP, EFABP, KFABP, PA-FABP, PAFABP
- Chromosome Location
- Chr. 8: 81280483 - 81284777 on Build GRCh38
- UniGene ID
- Hs.408061
- Species
- Homo sapiens
- Gene Symbol
- FABP5P3
- Entrez Gene ID
- 220832
- Gene Name
- fatty acid binding protein 5 pseudogene 3
- Gene Aliases
- FABP5L3
- Chromosome Location
- Chr. 7: 152436895 - 152443016 on Build GRCh38
- UniGene ID
- Hs.647108
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002935.1 |
|
2 |
462 |
|
GenBank mRNA |
BX338084.2 |
- |
2 |
462 |
|
View Full Product Details