Gene Transcripts
- Gene Symbol
- DOK1
- Entrez Gene ID
- 1796
- Gene Name
- docking protein 1
- Gene Aliases
- P62DOK, pp62
- Chromosome Location
- Chr. 2: 74549020 - 74557551 on Build GRCh38
- UniGene ID
- Hs.103854
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197260.1 |
NP_001184189.1
|
4, 5 |
740 |
|
NM_001318866.1 |
NP_001305795.1
|
4, 5 |
1240 |
|
NM_001318867.1 |
NP_001305796.1
|
4, 5 |
1285 |
|
NM_001318868.1 |
NP_001305797.1
|
3, 4 |
1057 |
|
NM_001318869.1 |
NP_001305798.1
|
3, 4 |
1051 |
|
NM_001381.4 |
NP_001372.1
|
4, 5 |
1242 |
|
GenBank mRNA |
AF035299.1 |
- |
4, 5 |
733 |
|
AF180527.1 |
- |
3, 4 |
462 |
|
AK055944.1 |
- |
4, 5 |
1242 |
|
AK222883.1 |
- |
4, 5 |
681 |
|
AK296154.1 |
- |
4, 5 |
695 |
|
AK300097.1 |
- |
3, 4 |
462 |
|
AK314678.1 |
- |
4, 5 |
662 |
|
BC111055.1 |
- |
3, 4 |
486 |
5752188 |
BC114440.1 |
- |
4, 5 |
668 |
40083624 |
BC114497.1 |
- |
3, 4 |
471 |
40083623 |
U70987.1 |
- |
4, 5 |
669 |
|
View Full Product Details