Gene Transcripts

Gene Symbol
Ofd1
Entrez Gene ID
237222
Gene Name
oral-facial-digital syndrome 1 gene homolog (human)
Gene Aliases
Cxorf5, DXGgc7e, ORF2
Chromosome Location
Chr. X: 166390033 - 166440838 on Build GRCm38
UniGene ID
Mm.247480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177429.3 NP_803178.2 4 748
GenBank mRNA AJ438159.1 - 4 599
BC119070.1 - 4 431 8733819
BC120487.1 - 4 431 40129410

View Full Product Details