Gene Transcripts

Gene Symbol
Fbxw13
Entrez Gene ID
211305
Gene Name
F-box and WD-40 domain protein 13
Gene Aliases
E330017E16
Chromosome Location
Chr. 9: 109179227 - 109195975 on Build GRCm38
UniGene ID
Mm.225242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177598.3 NP_808266.1 3 241
GenBank mRNA AK054339.1 - 3 242
BC119542.1 - 3 228 40056515

View Full Product Details