Gene Transcripts

Gene Symbol
Lrrtm3
Entrez Gene ID
216028
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
9630044H04Rik
Chromosome Location
Chr. 10: 63928497 - 64090255 on Build GRCm38
UniGene ID
Mm.431318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178678.4 NP_848793.1 3 2918
GenBank mRNA AK035882.1 - 3 2921
AK036180.1 - 3 2938
AK049844.1 - 1 1052
BC113178.1 - 3 2592 40060681
BC113784.1 - 3 2592 40060682

View Full Product Details