Gene Transcripts

Gene Symbol
Dpy19l3
Entrez Gene ID
233115
Gene Name
dpy-19-like 3 (C. elegans)
Gene Aliases
6030410G08, 9330164H19Rik
Chromosome Location
Chr. 7: 35685500 - 35754454 on Build GRCm38
UniGene ID
Mm.257808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178704.3 NP_848819.2 19 3131
GenBank mRNA AF538322.1 - 19 3115
AK033395.1 - 19 3088
AK034217.1 - 19 3132
AK045758.1 - 19 3032
BC050065.1 - 19 3075 30016211
BC059897.1 - 19 3027 6821433

View Full Product Details