Assay Design



Gene Transcripts

Gene Symbol
IAA34
Entrez Gene ID
838070
Gene Name
indole-3-acetic acid inducible 34
Gene Aliases
AT1G15050, T15D22.10, T15D22_10, indole-3-acetic acid inducible 34
Chromosome Location
Chr.1: 5181991 - 5183413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101375.3 NP_172959.2 1 - 2 258 88
GenBank mRNA AB493458.1 - 1 - 2 88 88
AF334713.1 - 1 - 2 88 88
AJ458327.1 - 1 - 2 88 88

View Full Product Details