Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084075.1 | NP_001077544.1 | 1 - 2 | 232 | 93 |
NM_001332191.1 | NP_001319013.1 | 1 - 2 | 289 | 93 | |
NM_001332192.1 | NP_001322095.1 | 1 - 2 | 232 | 93 | |
NM_001332193.1 | NP_001322096.1 | 1 - 2 | 232 | 93 | |
NM_001332194.1 | NP_001322097.1 | 1 - 2 | 232 | 93 | |
NM_101463.4 | NP_173047.1 | 1 - 2 | 232 | 93 | |
GenBank mRNA | AF321114.1 | - | 1 - 2 | 131 | 93 |
AF332459.1 | - | 1 - 2 | 131 | 93 | |
AK228419.1 | - | 1 - 2 | 230 | 93 | |
AY087316.1 | - | 1 - 2 | 233 | 93 | |
AY743921.1 | - | 1 - 2 | 131 | 93 |