Assay Design
Gene Transcripts
- Gene Symbol
- NRAMP6
- Entrez Gene ID
-
838166
- Gene Name
- NRAMP metal ion transporter 6
- Gene Aliases
-
AT1G15960, ATNRAMP6, NRAMP metal ion transporter 6, T24D18.6, T24D18_6
- Chromosome Location
-
Chr.1: 5481665 - 5486105 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101464.4 |
NP_173048.3 |
8 - 9 |
1799 |
105 |
GenBank mRNA |
AJ291831.1 |
- |
8 - 9 |
760 |
105 |
View Full Product Details