Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035979.1 | NP_001031056.1 | 2 - 3 | 344 | 126 |
NM_001198083.1 | NP_001185012.1 | 3 - 4 | 417 | 126 | |
NM_001332226.1 | NP_001322506.1 | 2 - 3 | 328 | 126 | |
NM_001332227.1 | NP_001322507.1 | 2 - 3 | 338 | 126 | |
NM_101511.6 | NP_563998.1 | 2 - 3 | 388 | 126 | |
NM_179344.2 | NP_849675.1 | 2 - 3 | 328 | 126 | |
GenBank mRNA | AB032865.1 | - | 2 - 3 | 250 | 126 |
AJ010500.1 | - | 2 - 3 | 258 | 126 | |
AK229282.1 | - | 2 - 3 | 347 | 126 | |
AY084558.1 | - | 2 - 3 | 328 | 126 | |
BT024876.1 | - | 1 - 2 | 185 | 126 |