Assay Design



Gene Transcripts

Gene Symbol
KU70
Entrez Gene ID
838268
Gene Name
ATP-dependent DNA helicase 2 subunit Ku70-like protein
Gene Aliases
ARABIDOPSIS THALIANA KU70 HOMOLOG, AT1G16970, ATKU70, F6I1.2, F6I1_2, KU70 homolog, KU70-LIKE PROTEIN
Chromosome Location
Chr.1: 5800767 - 5805811 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101558.4 NP_564012.1 10 - 11 1092 123
GenBank mRNA AF283759.1 - 10 - 11 1055 123
AK221642.1 - 10 - 11 1080 123

View Full Product Details