Assay Design



Gene Transcripts

Gene Symbol
TPS2
Entrez Gene ID
838269
Gene Name
trehalose-6-phosphate synthase
Gene Aliases
AT1G16980, ATTPS2, F6I1.1, F6I1_1, TREHALOSE -6-PHOSPHATASE SYNTHASE S2, trehalose-phosphatase/synthase 2
Chromosome Location
Chr.1: 5807200 - 5811610 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332273.1 NP_001321461.1 7 - 8 1194 102
NM_101559.2 NP_173142.2 7 - 8 1194 102

View Full Product Details