Assay Design



Gene Transcripts

Gene Symbol
CYP72C1
Entrez Gene ID
838276
Gene Name
cytochrome p450 72c1
Gene Aliases
AT1G17060, CHI2, CHIBI 2, F20D23.24, F20D23_24, SHK1, SHRINK 1, SOB7, SUPPRESSOR OF PHYB-4 7, cytochrome p450 72c1
Chromosome Location
Chr.1: 5832094 - 5835488 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332275.1 NP_001319024.1 3 - 4 975 85

View Full Product Details