Assay Design
Gene Transcripts
- Gene Symbol
- CYP72C1
- Entrez Gene ID
-
838276
- Gene Name
- cytochrome p450 72c1
- Gene Aliases
-
AT1G17060, CHI2, CHIBI 2, F20D23.24, F20D23_24, SHK1, SHRINK 1, SOB7, SUPPRESSOR OF PHYB-4 7, cytochrome p450 72c1
- Chromosome Location
-
Chr.1: 5832094 - 5835488 on Build TAIR10.1
View Full Product Details