Assay Design



Gene Transcripts

Gene Symbol
GSTU26
Entrez Gene ID
838290
Gene Name
glutathione S-transferase tau 26
Gene Aliases
AT1G17190, ATGSTU26, F20D23.11, F20D23_11, glutathione S-transferase tau 26
Chromosome Location
Chr.1: 5875295 - 5876620 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101580.4 NP_173162.1 1 - 2 478 73
GenBank mRNA AJ306688.1 - 1 - 2 363 73
AK227997.1 - 1 - 2 399 73
BT004605.1 - 1 - 2 321 73

View Full Product Details