Assay Design



Gene Transcripts

Gene Symbol
UBP1B
Entrez Gene ID
838309
Gene Name
oligouridylate binding protein 1B
Gene Aliases
AT1G17370, F28G4.17, oligouridylate binding protein 1B
Chromosome Location
Chr.1: 5951542 - 5955143 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123829.1 NP_001117301.1 1 - 2 313 73
NM_101598.3 NP_564018.1 1 - 2 419 73
GenBank mRNA AY085705.1 - 1 - 2 221 73
BT026477.1 - 1 - 2 101 73

View Full Product Details