Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_101634.4 | NP_564033.1 | 10 - 11 | 1510 | 96 |
NM_179350.4 | NP_849681.1 | 10 - 11 | 1511 | 96 | |
GenBank mRNA | AF370519.1 | - | 10 - 11 | 1401 | 96 |
AK226540.1 | - | 10 - 11 | 1454 | 96 | |
AY045665.1 | - | 10 - 11 | 1401 | 96 | |
BT000645.1 | - | 10 - 11 | 1136 | 96 | |
BT002183.1 | - | 10 - 11 | 1133 | 96 | |
U40161.1 | - | 10 - 11 | 1222 | 96 |