Assay Design
Gene Transcripts
- Gene Symbol
- MYB52
- Entrez Gene ID
-
838374
- Gene Name
- myb domain protein 52
- Gene Aliases
-
AT1G17950, ATMYB52, BW52, F2H15.17, F2H15_17, MYB DOMAIN PROTEIN 52, myb domain protein 52
- Chromosome Location
-
Chr.1: 6177481 - 6179289 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101658.3 |
NP_173237.1 |
2 - 3 |
832 |
82 |
GenBank mRNA |
AY519554.1 |
- |
2 - 3 |
549 |
82 |
View Full Product Details