Assay Design



Gene Transcripts

Gene Symbol
PSP
Entrez Gene ID
838445
Gene Name
3-phosphoserine phosphatase
Gene Aliases
3-PHOSPHOSERINE PHOSPHATASE, 3-phosphoserine phosphatase, AT1G18640, PSP1, phosphoserine phosphatase 1
Chromosome Location
Chr.1: 6416169 - 6418693 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332354.1 NP_001320464.1 2 - 3 359 83
NM_202129.3 NP_973858.1 2 - 3 362 83
GenBank mRNA AY065351.1 - 2 - 3 353 83
AY087385.1 - 2 - 3 314 83
AY096687.1 - 1 - 2 216 83

View Full Product Details