Assay Design



Gene Transcripts

Gene Symbol
ARF19
Entrez Gene ID
838505
Gene Name
auxin response factor 19
Gene Aliases
ARF11, AT1G19220, AUXIN RESPONSE FACTOR11, IAA22, T29M8.9, T29M8_9, auxin response factor 19, indole-3-acetic acid inducible 22
Chromosome Location
Chr.1: 6627683 - 6633133 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101780.4 NP_173356.1 9 - 10 1495 129
GenBank mRNA AB493467.1 - 9 - 10 1141 129
AY669794.1 - 9 - 10 1141 129
U53672.2 - 9 - 10 1359 129

View Full Product Details