Assay Design
Gene Transcripts
- Gene Symbol
- NFD5
- Entrez Gene ID
-
838538
- Gene Name
- pentatricopeptide (PPR) repeat-containing protein
- Gene Aliases
-
AT1G19520, F18O14.40, NUCLEAR FUSION DEFECTIVE 5
- Chromosome Location
-
Chr.1: 6759859 - 6762847 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101809.4 |
NP_564088.2 |
2 - 2 |
2304 |
73 |
View Full Product Details