Assay Design



Gene Transcripts

Gene Symbol
MAPKKK5
Entrez Gene ID
836819
Gene Name
mitogen-activated protein kinase kinase kinase 5
Gene Aliases
AT5G66850, MUD21.11, MUD21_11, mitogen-activated protein kinase kinase kinase 5
Chromosome Location
Chr.5: 26695568 - 26699426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345806.1 NP_001330515.1 6 - 7 1760 77
NM_126084.3 NP_569040.1 6 - 7 1760 77
GenBank mRNA AF360242.1 - 6 - 7 1658 77
AY040040.1 - 6 - 7 1493 77

View Full Product Details