Assay Design



Gene Transcripts

Gene Symbol
ICU2
Entrez Gene ID
836845
Gene Name
DNA-directed DNA polymerase
Gene Aliases
AT5G67100, INCURVATA2, K21H1.14, K21H1_14
Chromosome Location
Chr.5: 26776953 - 26786431 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345811.1 NP_001331725.1 14 - 15 2908 122
NM_126110.3 NP_201511.2 14 - 15 2890 122

View Full Product Details