Assay Design



Gene Transcripts

Gene Symbol
CKA1
Entrez Gene ID
836873
Gene Name
casein kinase alpha 1
Gene Aliases
AT5G67380, ATCKA1, ATPK12D, K8K14.10, K8K14_10, casein kinase alpha 1
Chromosome Location
Chr.5: 26880966 - 26884079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037088.2 NP_001032165.1 6 - 7 891 124
NM_126138.4 NP_201539.2 6 - 7 1649 124
GenBank mRNA AK317458.1 - 6 - 7 893 124

View Full Product Details