Assay Design
Gene Transcripts
- Gene Symbol
- DG1
- Entrez Gene ID
-
836893
- Gene Name
- Tetratricopeptide repeat (TPR)-like superfamily protein
- Gene Aliases
-
AT5G67570, DELAYED GREENING 1, EMB1408, EMB246, EMBRYO DEFECTIVE 246, K9I9.14, K9I9_14, embryo defective 1408
- Chromosome Location
-
Chr.5: 26952341 - 26955543 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_126157.4 |
NP_201558.3 |
4 - 5 |
1051 |
77 |
View Full Product Details