Assay Design



Gene Transcripts

Gene Symbol
STL2P
Entrez Gene ID
814675
Gene Name
SEC12P-like 2 protein
Gene Aliases
AT2G01470, ATSEC12, F2I9.9, F2I9_9, SEC12P-like 2 protein
Chromosome Location
Chr.2: 211913 - 214473 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126208.4 NP_178256.1 6 - 7 820 100
GenBank mRNA AY074332.1 - 6 - 7 811 100
AY123030.1 - 6 - 7 782 100

View Full Product Details