Assay Design



Gene Transcripts

Gene Symbol
PAP7
Entrez Gene ID
814719
Gene Name
purple acid phosphatase 7
Gene Aliases
AT2G01880, ATPAP7, PURPLE ACID PHOSPHATASE 7, T23K3.7, T23K3_7, purple acid phosphatase 7
Chromosome Location
Chr.2: 391421 - 394184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126249.5 NP_178297.2 6 - 7 994 66
GenBank mRNA AF492659.1 - 6 - 7 777 66
BT026449.1 - 6 - 7 777 66

View Full Product Details