Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_126341.3 | NP_178389.1 | 7 - 8 | 1045 | 75 |
NM_201675.2 | NP_973404.1 | 7 - 8 | 1020 | 75 | |
GenBank mRNA | AJ289165.1 | - | 7 - 8 | 722 | 75 |
AK226970.1 | - | 7 - 8 | 849 | 75 |