Assay Design



Gene Transcripts

Gene Symbol
EXPA15
Entrez Gene ID
814838
Gene Name
expansin A15
Gene Aliases
AT2G03090, ATEXP15, ATEXPA15, ATHEXP ALPHA 1.3, EXP15, EXPANSIN 15, T17M13.26, T17M13_26, expansin A15
Chromosome Location
Chr.2: 916806 - 918671 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126361.4 NP_178409.2 2 - 3 256 91
GenBank mRNA AY072167.1 - 2 - 3 228 91
AY117342.1 - 1 - 2 139 91
AY510271.1 - 2 - 3 214 91

View Full Product Details