Assay Design



Gene Transcripts

Gene Symbol
VMA10
Entrez Gene ID
821130
Gene Name
vacuolar membrane ATPase 10
Gene Aliases
AT3G01390, AVMA10, T13O15.3, T13O15_3, VACUOLAR MEMBRANE ATPASE SUBUNIT G, VHA-G1, vacuolar membrane ATPase 10
Chromosome Location
Chr.3: 149940 - 151572 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337333.1 NP_001327895.1 3 - 4 720 82
NM_001337334.1 NP_001327894.1 3 - 4 590 82
NM_111005.6 NP_186788.1 3 - 4 296 82
NM_180158.3 NP_850489.1 2 - 3 760 82
GenBank mRNA AF181688.1 - 2 - 3 224 82
AJ005901.1 - 2 - 3 220 82
AY072365.1 - 3 - 4 253 82
AY114730.1 - 2 - 3 182 82

View Full Product Details