Assay Design



Gene Transcripts

Gene Symbol
DOX1
Entrez Gene ID
821135
Gene Name
Peroxidase superfamily protein
Gene Aliases
ALPHA-DOX1, AT3G01420, DIOX1, PADOX-1, T13O15.6, T13O15_6, alpha-dioxygenase 1, plant alpha dioxygenase 1
Chromosome Location
Chr.3: 159450 - 162905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111008.3 NP_186791.1 1 - 2 289 68
GenBank mRNA AY042787.1 - 1 - 2 221 68
AY064666.1 - 1 - 2 110 68

View Full Product Details