Assay Design



Gene Transcripts

Gene Symbol
HB-1
Entrez Gene ID
821138
Gene Name
homeobox 1
Gene Aliases
ARABIDOPSIS THALIANA HOMEOBOX 1, AT3G01470, ATHB-1, ATHB1, F4P13.2, F4P13_2, HAT5, HD-ZIP-1, HOMEODOMAIN PROTEIN FROM ARABIDOIPSIS THALIANA 5, homeobox 1
Chromosome Location
Chr.3: 182396 - 184403 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111013.4 NP_186796.1 2 - 3 604 64
GenBank mRNA AY058188.1 - 2 - 3 604 64
AY098972.1 - 2 - 3 235 64
X58821.1 - - 565 64

View Full Product Details