Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001337342.1 | NP_001326777.1 | 2 - 3 | 776 | 67 |
NM_111016.4 | NP_186799.2 | 2 - 3 | 568 | 67 | |
NM_180159.2 | NP_850490.1 | 2 - 3 | 380 | 67 | |
NM_180160.4 | NP_850491.1 | 2 - 3 | 1072 | 67 | |
GenBank mRNA | AF428284.1 | - | 2 - 3 | 332 | 67 |
AF428459.1 | - | 2 - 3 | 382 | 67 | |
AK226447.1 | - | 2 - 3 | 323 | 67 | |
AY056175.1 | - | 2 - 3 | 332 | 67 | |
AY062785.1 | - | 2 - 3 | 299 | 67 | |
AY081658.1 | - | 2 - 3 | 281 | 67 | |
AY091066.1 | - | 2 - 3 | 377 | 67 | |
X65541.1 | - | - | 368 | 67 |