Assay Design



Gene Transcripts

Gene Symbol
MYB57
Entrez Gene ID
821113
Gene Name
myb domain protein 57
Gene Aliases
AT3G01530, ATMYB57, F4P13.8, F4P13_8, myb domain protein 57
Chromosome Location
Chr.3: 210119 - 211804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111019.2 NP_186802.1 1 - 2 827 73
GenBank mRNA AK118091.1 - 1 - 2 282 73
AY519582.1 - 1 - 2 174 73
BT005574.1 - 1 - 2 174 73

View Full Product Details