Assay Design



Gene Transcripts

Gene Symbol
SEP2
Entrez Gene ID
821151
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 4, AGL4, AT3G02310, F14P3.4, F14P3_4, SEPALLATA 2, TRANSCRIPTION FACTOR
Chromosome Location
Chr.3: 464279 - 467151 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111098.4 NP_186880.1 5 - 6 977 74
GenBank mRNA BT020478.1 - 5 - 6 525 74
M55552.1 - 5 - 6 851 74

View Full Product Details