Assay Design
Gene Transcripts
- Gene Symbol
- XPO1B
- Entrez Gene ID
-
821074
- Gene Name
- exportin 1B
- Gene Aliases
-
AT3G03110, CRM1B, T17B22.20, T17B22_20, exportin 1B
- Chromosome Location
-
Chr.3: 708698 - 717093 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111181.3 |
NP_566193.2 |
13 - 14 |
1436 |
65 |
View Full Product Details