Assay Design



Gene Transcripts

Gene Symbol
DCL2
Entrez Gene ID
821300
Gene Name
dicer-like 2
Gene Aliases
AT3G03300, ATDCL2, DICER-LIKE 2, T17B22.28, dicer-like 2
Chromosome Location
Chr.3: 767880 - 776214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084632.1 NP_001078101.1 19 - 20 3795 90
NM_001202869.2 NP_001189798.1 22 - 23 4158 90
NM_001337464.1 NP_001327050.1 16 - 17 3276 90
NM_111200.5 NP_566199.4 23 - 24 4400 90

View Full Product Details