Assay Design



Gene Transcripts

Gene Symbol
PEX19-1
Entrez Gene ID
821248
Gene Name
peroxin 19-1
Gene Aliases
AT3G03490, AtPEX19-1, T21P5.9, T21P5_9, peroxin 19-1
Chromosome Location
Chr.3: 830116 - 831997 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111220.4 NP_186999.1 4 - 5 941 57
GenBank mRNA AJ564199.1 - 4 - 5 808 57
BT012633.1 - 4 - 5 703 57

View Full Product Details