Assay Design
Gene Transcripts
- Gene Symbol
- RIF10
- Entrez Gene ID
-
821181
- Gene Name
- polyribonucleotide nucleotidyltransferase
- Gene Aliases
-
AT3G03710, F20H23.26, PDE326, PIGMENT DEFECTIVE 326, PNP, POLYNUCLEOTIDE PHOSPHORYLASE, resistant to inhibition with FSM 10
- Chromosome Location
-
Chr.3: 919362 - 925372 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111242.3 |
NP_187021.2 |
11 - 12 |
1508 |
132 |
GenBank mRNA |
AF450480.1 |
- |
11 - 12 |
1328 |
132 |
View Full Product Details