Assay Design



Gene Transcripts

Gene Symbol
AK-HSDH II
Entrez Gene ID
827715
Gene Name
aspartate kinase-homoserine dehydrogenase ii
Gene Aliases
AK-HSDH, ASPARTATE KINASE-HOMOSERINE DEHYDROGENASE, AT4G19710, T16H5.70, T16H5_70, aspartate kinase-homoserine dehydrogenase ii
Chromosome Location
Chr.4: 10724814 - 10729768 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118091.3 NP_193706.2 12 - 13 1991 100
NM_202847.3 NP_974576.1 12 - 13 2032 100
GenBank mRNA AK228252.1 - 12 - 13 1960 100

View Full Product Details