Assay Design



Gene Transcripts

Gene Symbol
PCAP1
Entrez Gene ID
827773
Gene Name
plasma-membrane associated cation-binding protein 1
Gene Aliases
ARABIDOPSIS THALIANA PLASMA-MEMBRANE ASSOCIATED CATION-BINDING PROTEIN 1, AT4G20260, ATPCAP1, F1C12.180, F1C12_180, MDP25, microtubule-destabilizing protein 25, plasma-membrane associated cation-binding protein 1
Chromosome Location
Chr.4: 10940414 - 10943561 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036599.2 NP_001031676.1 4 - 5 389 69
NM_001036600.2 NP_001031677.1 4 - 5 411 69
NM_001203849.1 NP_001190778.1 4 - 5 408 69
NM_001203850.1 NP_001190779.1 4 - 5 408 69
NM_001341407.1 NP_001320005.1 4 - 5 740 69
NM_001341408.1 NP_001329494.1 4 - 5 938 69
NM_001341409.1 NP_001329493.1 4 - 5 624 69
NM_001341410.1 NP_001329492.1 4 - 5 601 69
NM_118145.4 NP_193759.1 4 - 5 379 69
NM_179081.5 NP_849412.1 4 - 5 417 69
GenBank mRNA AK317524.1 - 4 - 5 375 69
AY093084.1 - 4 - 5 408 69
AY128768.1 - 3 - 4 282 69
Y08061.1 - - 410 69

View Full Product Details